missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metolazone, Fisher BioReagents
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Packaging:
Amber Glass
Unit Size:
100mg
25mg
50mg
Description
Metolazone is an anti-hypertensive.
Specifications
Specifications
| Melting Point | 248°C |
| Color | White |
| Packaging | Amber Glass |
| Quantity | 50 mg |
| Merck Index | 15, 6224 |
| IUPAC Name | 7-chloro-2-methyl-3-(2-methylphenyl)-4-oxo-1,2,3,4-tetrahydroquinazoline-6-sulfonamide |
| Formula Weight | 365.8 |
| Physical Form | Powder or Solid |
| Chemical Name or Material | Metolazone |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction