missing translation for 'onlineSavingsMsg'
Learn More

Metro™ Cage Rack Lab Cart, Autoclavable

Preconfigured cage racks available in two footprints

Brand:  Metro™ LC5S1860

Additional Details : Weight : 49.35200kg

 View more versions of this product

Product Code. 15987011

  • £3600.00 / Each
Estimated Shipment: 09-10-2024
to see stock.
Add to basket

Description

Description

  • Carts come standard with swedged posts and aluminum split sleeves
  • Units measure 69 in. high (175.3cm) including casters
  • Footprints: 18 x 60 in. (45.7 x 152.4cm) or 24 x 60 in. (61 x 152.4cm)

Order Info

Casters must be ordered separately.

Specifications

Specifications

Cage Rack
Ideal solution for hi-temp, washable applications
175.3 cm
60 in.
Stainless Steel
18 in.
60 x 18 x 69 in. (152.4 x 45.7 x 175.3 cm)
69 in.
5 Stainless Steel autoclave shelves with swedged posts and Aluminum split sleeves
152.4 cm
5
45.7 cm
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions