Learn More
Metro™ Cover Holder for Super Erecta Wire Shelves
Accessory for Super Erecta Wire Shelf
Brand:  Metro™ LAR-CH
Product Code. 15946971
  • £120.00 / Each
Estimated Shipment: 24-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Holds tote box covers
  • Stainless steel wire
  • 18.25L x 18.5W x 4 in. H;
  • 6 lbs.
Specifications
Specifications
Cover Holder
Metro™ Super Erecta™ shelving system
10.16 cm
46.4 cm
2.7 kg
47 cm
18.25 x 18.5 x 4 in. (46.4 x 47 x 10.16 cm)
4 in.
18.25 in.
6 lb.
18.5 in.
Stainless Steel