missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metro™ Flexline™ Cart Accessories
Side/recessed storage accessories
Brand: Metro™ FL252
This item is not returnable.
View return policy
Description
- Fits Metro FL251 sharps containter
- 20 5-quart containters
Specifications
51.25 gal. | |
194 L | |
13 x 4.75 x 11.87 in. (33.02 x 12.06 x 30.14 cm) | |
11.87 in. | |
13 in. | |
Plastic | |
12.06 cm |
Replacement Lockable Sharps Containers | |
Red | |
For cart | |
30.14 cm | |
33.02 cm | |
4.75 in. |