Learn More
Metro™ HD Super Stainless Lab Worktable, Stationary
Keeping things where the belong in a lab is important to all staff. The Metro HD Super Stainless Lab Worktable is stationary and features an open base that makes it easy to clean. Its stable surface and base construction makes it a choice fit for nearly any lab.
£1560.00 - £3905.00
Product Code | Brand | Dimensions (L x W x H) | Material | Includes | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W x H) | Material | Includes | Price | Quantity & Availability | |||
15967331
|
Metro™
WT366US |
60 x 36 x 34 in. (152.4 x 91.4 x 86.4 cm) | Stainless Steel | Bottom 3-sided frame (SS) |
£1975.00
Each |
Estimated Shipment: 06-09-2024
Log in to see stock. |
|||
15977331
|
Metro™
WT367US |
72 x 36 x 34 in. (91.4 x 91.4 x 86.36 cm) | Stainless Steel | Bottom 3-sided frame (SS) |
£2140.00
Each |
Estimated Shipment: 06-09-2024
Log in to see stock. |
|||
15927331
|
Metro™
WT307US |
70 x 30 x 34 in. (177.8 x 76.2 x 86.36 cm) | Stainless Steel | Bottom 3-sided frame (SS) |
£1845.00
Each |
Estimated Shipment: 06-09-2024
Log in to see stock. |
|||
15957321
|
Metro™
WT305FS |
48 x 30 x 34 in. (121.92 x 76.2 x 86.36 cm) | Stainless Steel (14 ga.) | Bottom shelf (SS) |
£1840.00
Each |
Estimated Shipment: 06-09-2024
Log in to see stock. |
|||
15967321
|
Metro™
WT305US |
48 x 30 x 34 in. (121.92 x 76.2 x 86.36 cm) | Stainless Steel (14 ga.) | Bottom 3-sided frame (SS) |
£1565.00
Each |
Estimated Shipment: 06-09-2024
Log in to see stock. |
|||
15977321
|
Metro™
WT306FS |
60 x 30 x 34 in. (152.4 x 76.2 x 86.36 cm) | Stainless Steel (14 ga.) | Bottom shelf (SS) |
£1960.00
Each |
Estimated Shipment: 06-09-2024
Log in to see stock. |
|||
15907331
|
Metro™
WT307FC |
72 x 30 x 34 in. (182.88 x 76.2 x 86.36 cm) | Stainless Steel (14 ga.) | Bottom shelf (galvanized) |
£1560.00
Each |
Estimated Shipment: 06-09-2024
Log in to see stock. |
|||
15987321
|
Metro™
WT306HS |
60 x 30 x 34 in. (152.4 x 76.2 x 86.36 cm) | Stainless Steel (14 ga.) | Bottom H-frame (SS) |
£1695.00
Each |
Estimated Shipment: 06-09-2024
Log in to see stock. |
|||
15917331
|
Metro™
WT307FS |
72 x 30 x 34 in. (182.88 x 76.2 x 86.36 cm) | Stainless Steel (14 ga.) | Bottom shelf (SS) |
£2125.00
Each |
Estimated Shipment: 06-09-2024
Log in to see stock. |
|||
15937331
|
Metro™
WT309FC |
96 x 30 x 34 in. (243.84 x 76.2 x 86.36 cm) | Stainless Steel (14 ga.) | Bottom shelf (galvanized) |
£1870.00
Each |
Estimated Shipment: 06-09-2024
Log in to see stock. |
|||
15947331
|
Metro™
WT309FS |
96 x 30 x 34 in. (243.84 x 76.2 x 86.36 cm) | Stainless Steel (14 ga.) | Bottom shelf (SS) |
£3905.00
Each |
Estimated Shipment: 18-09-2024
Log in to see stock. |
|||
15957331
|
Metro™
WT366FC |
60 x 36 x 34 in. (152.4 x 91.44 x 86.36 cm) | Stainless Steel (14 ga.) | Bottom shelf (galvanized) |
£1785.00
Each |
Estimated Shipment: 06-09-2024
Log in to see stock. |
|||
15997321
|
Metro™
WT306US |
60 x 30 x 34 in. (152.5 x 76.2 x 86.36 cm) | Stainless Steel | Bottom 3-sided frame (SS) |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
- 34 in. high (86.4cm) seamless, 14-gauge stainless-steel work surface with rounded edges
- Support structure virtually eliminates “drumming”
- Posts with adjustable feet compensate for slight floor irregularities
- Leveling foot on each post
Order Info
Shipped knocked down. Assemble easily without tools. Other sizes available; call your Fisher Safety Customer Service Representative at 1-800-772-6733.
Specifications
34 in. | |
Stationary Table |