missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metro™ MW Series Utility Carts with Solid Shelves
Move and store items more conveniently in any laboratory. Metro MW Series Utility Carts are constructed of Super Erecta™ shelves with plastic split sleeves, two one-piece handles and casters with donut bumpers to make mobility of lab equipment easy.
£1345.00 - £1610.00
Specifications
Product Type | Utility Cart |
---|---|
Certifications/Compliance | NSF |
Description | 3-tier Model |
For Use With (Application) | Transporting materials that require an expanse of flat, solid surface |
Load Bearing Capacity (English) | 375 lb. |
Product Code | Brand | Height (English) | Width (English) | Length (English) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (English) | Width (English) | Length (English) | Price | Quantity & Availability | |||
15947171
|
Metro™
MW203 |
38 in. | 18 in. | 24 in. |
£1345.00
Each |
Estimated Shipment: 26-09-2024
Log in to see stock. |
|||
15957171
|
Metro™
MW206 |
39 in. | 21 in. | 36 in. |
£1610.00
Each |
Estimated Shipment: 26-09-2024
Log in to see stock. |
|||
Description
Cart
- Load capacity: 375 lb. (170.1kg)
- Fully interchangeable Super Erecta shelves
- Center shelf can be positioned at any height in 1 in. (2.5cm) increments
- One-piece stainless-steel handles add rigidity and durability
- 18 in. (46cm) carts have 4 in. dia. (10cm) casters; 21 in. (53cm) and 24 in. (61cm) wide carts have 5 in. dia. (13cm) casters
- Three tiers
- 375 lbs (170kg) weight capacity per cart
- 18-gauge stainless-steel, doubly thick at the edges for rigidity and strength
- Raised edges help contain spills and make cleanup easier
- Sturdy corner castings facilitate mounting
Order Info
Shipped knocked down. Assembles/disassembles quickly and easily without tools.
Specifications
Utility Cart | |
3-tier Model | |
375 lb. | |
3 |
NSF | |
Transporting materials that require an expanse of flat, solid surface | |
170 kg |