missing translation for 'onlineSavingsMsg'
Learn More

Metro™ Stainless Lab Worktable Accessory, Keyboard Tray

For use with Metro Stainless Steel Worktables

Brand:  Metro™ LTSKB

Product Code. 15937121

  • £494.00 / Each
Estimated Shipment: 26-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Stainless steel
  • Fits 48 and 60 in. (122 and 152cm) table lengths

Specifications

Specifications

48 and 60 in. Stainless Steel mobile table
Stainless Steel mobile table
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions