Learn More
Metro™ Stainless Lab Worktable, Black Phenolic Top and 3-Sided Frame
Protect your workers in the laboratory. Metro Stainless Lab Worktable helps to keep them safe with a phenolic top and corrosion-proof, stainless steel construction throughout the body.
Brand: Metro™ LTSM36UPB
Additional Details : Weight : 29.75200kg
Description
- Easily cleaned smooth surfaces
- Mobile units can be easily moved to clean behind table area
- Wide range of accessories and caster options available
Order Info
Casters must be ordered separately.
Specifications
35.75 in. | |
35.75 x 30 x 35.8 in. (90.8 x 76.2 x 91.1 cm) | |
91.1 cm | |
3 Sided frame, adaptor insert | |
273 kg | |
90.8 cm | |
Stainless Steel |
Mobile Table | |
Ideal for lab | |
35.8 in. | |
30 in. | |
76.2 cm | |
600 lb. |