missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metro™ Stainless Lab Worktable, Gray Phenolic Top and Solid HD Shelf
Metro Stainless Lab Worktable features corrosion-proof performance and stainless steel construction throughout.
Brand: Metro™ LTS48PG
This item is not returnable.
View return policy
Description
- Easily cleaned smooth surfaces
- Mobile units can be easily moved to clean behind table area
- Wide range of accessories and caster options available
Order Info
Casters must be ordered separately.
Specifications
Worktable | |
Ideal for lab | |
91.44 cm | |
47.75 in. | |
600 lb. | |
113 lb. | |
30 in. | |
Stainless Steel |
47.75 x 30 x 36 in. | |
36 in. | |
Solid HD shelf | |
121.2 cm | |
273 kg | |
54 kg | |
76.2 cm |