missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metro™ Stainless Lab Worktable, Stainless Top, With Backsplash and 3-Sided Frame
Ensure your workers remain safe in the laboratory. Metro Stainless Lab Worktable with backsplash, 3-sided frame and stainless steel construction throughout provides cleanliness along with corrosion-proof performance.
Brand: Metro™ LTS36US
This item is not returnable.
View return policy
Description
- Easily cleaned smooth surfaces
- Mobile units can be easily moved to clean behind table area
- Wide range of accessories and caster options available
Order Info
Casters must be ordered separately.
Specifications
Stationary Table | |
Ideal for lab | |
89 cm | |
35.75 in. | |
600 lb. | |
30 in. | |
Stainless Steel |
35.75 x 30 x 35 in. | |
35 in. | |
Backsplash with 3 sided frame | |
90.8 cm | |
273 kg | |
76.2 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction