Learn More
Metro™ Stainless Lab Worktable, Stainless Top, With Backsplash and 3-Sided Frame
Ensure your workers remain safe in the laboratory. Metro Stainless Lab Worktable with backsplash, 3-sided frame and stainless steel construction throughout provides cleanliness along with corrosion-proof performance.
Brand:  Metro™ LTS30US
 View more versions of this product
Product Code. 15977091
  • £1730.00 / Each
Estimated Shipment: 26-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Easily cleaned smooth surfaces
  • Mobile units can be easily moved to clean behind table area
  • Wide range of accessories and caster options available
Order Info
Casters must be ordered separately.
Specifications
Specifications
Stationary Table
Ideal for lab
89 cm
29.75 in.
600 lb.
30 in.
Stainless Steel
29.75 x 30 x 35 in.
35 in.
Backsplash with 3 sided frame
75.5 cm
273 kg
76.2 cm