missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metro™ Stainless Laboratory Worktable Accessory, Drawer
Stow items away safely and neatly with a drawer add-on accessory. Metrol Stainless Laboratory Worktable Drawer comes in sizes for 30, 36, 48, or 60 in. (76, 91, 122, and 152cm) table lengths.
£175.00 - £975.00
Specifications
For Use With (Equipment) | Stationary or mobile 30, 36, 48 and 60 in. W worktables |
---|
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
15927121
|
Metro™
LTSD6 |
Regular Drawer |
£499.00
Each |
Estimated Shipment: 26-09-2024
Log in to see stock. |
|||||
15987341
|
Metro™
WTD51S |
Deluxe Drawer |
£975.00
Each |
Estimated Shipment: 26-09-2024
Log in to see stock. |
|||||
15977341
|
Metro™
WTD21C |
Economy Drawer |
£175.00
Each |
Estimated Shipment: 26-09-2024
Log in to see stock. |
|||||
Description
- Can be used with stationary or mobile worktablesl
Specifications
Stationary or mobile 30, 36, 48 and 60 in. W worktables |