missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metro™ Standard-Duty Mobile Security Units, Stainless Steel
Standard-duty mobile security units provide protection of valuable materials and sensitive items from loss or pilferage. Available in a sleek, easy-to-clean stainless steel finish.
Brand: Metro™ SEC63S-SD
This item is not returnable.
View return policy
Description
- Mobile security units provide protection for valuables and sensitive materials from loss or pilferage.
- Type 304 Stainless provides superior corrosion resistance.
- Heavy gauge open wire construction keeps contents visible at all times, making it easy to check inventory.
- Choice of casters to match requirements.
- Patented, ergonomic, 1/4-turn door handle.
- Double doors open 270° and can be secured along the sides of the unit.
- Fixed top and bottom Super Erecta shelves.
- Optional intermediate shelves can be postitioned in 1 in. increments along entire height of post.
Order Info
Shipped knocked down; assemble easily. Super Adjustable/Super Erecta™ intermediate shelves and casters sold separately.
Specifications
Mobile Security Cart | |
Silver | |
62 in. | |
40.75 in. | |
Stainless Steel | |
76 kg | |
33.5 in. |
70 kg | |
40.75 x 33.5 x 62 in. (103.5 x 85 x 157.5 cm) | |
157.5 cm | |
103.5 cm | |
2 | |
167 lb. | |
85 cm |