Learn More
Metro™ Starsys™ Overhead Storage Unit
Microban™ antimicrobial protection prevents growth of stains and odor-causing bacteria.
Brand:  Metro™ SXRDOH27P2C
 View more versions of this product
Product Code. 15987241
  • £3290.00 / Each
Explore more special offers
Description
Description
  • Built on same widths as Starsys base units utilizing very similar construction
  • Insures that wall cabinets align with lower product configuration in application
  • All overhead cabinets include corrugated polymer inner panels in their assembly as well as clean-design back panel, molded with cosmetic side facing inside of cabinet, creating much cleaner appearance - especially when used without doors or in conjunction with clear door
  • Necessary wall mount rails/hardware are included with each wall cabinet
  • Microban™ antimicrobial protection inhibits growth of stain and odor-causing bacteria on Starsys, keeping surface areas cleaner between cleanings
Specifications
Specifications
ISO 9001-9002
Fixed Window
Doors with View, Double Width, Solid Shelves
2
41.3 in.
13.8 in.
Polymer
41.3 in.
13.8 x 41.3 x 29.5 in.
29.5 in.
2
Solid Shelf
Taupe
29.5 in.
Overhead Storage Unit