missing translation for 'onlineSavingsMsg'
Learn More

Metro™ Starsys™ Preconfigured Mobile Workstation, General Supply Cart

Made from an easy-to-clean, advanced polymer material that will not dent, chip, rust, flake, or corrode like metal carts

Brand:  Metro™ SXRS43CM4

Additional Details : Weight : 72.57600kg

Product Code. 15987251

  • £5900.00 / Each
Estimated Shipment: 08-08-2024
to see stock.
Add to basket

Includes: Mobile shell with mechanical shell with mechanical keylock, 3 in. Polymer drawer with locking cover, three 3 in. Polymer drawer, two 6 in. Polymer drawer, 9 in. Polymer drawer two 3 in. drawer divider kits, taupe drawer pull, left swingout pod locking til

Description

Description

  • Microban™ antimicrobial protection inhibits the growth of stain- and odor-causing bacteria
  • Carts utilize 5 in. (127mm) polyurethane casters: two front casters with easily accessed toe brakes and two rear swivel casters
  • Each drawer face has a 0.5 × 18.38 in. (1.3 × 46.7cm) polished area allowing for secure adhesion of DYMO™ and Zebra™ labels (not included)

Specifications

Specifications

Preconfigured Cart
110 cm
24.9 in.
Mobile shell with mechanical shell with mechanical keylock, 3 in. Polymer drawer with locking cover, three 3 in. Polymer drawer, two 6 in. Polymer drawer, 9 in. Polymer drawer two 3 in. drawer divider kits, taupe drawer pull, left swingout pod locking tilt bins, waste basket and holder
22.8 in.
44.7 in.
Epoxy-coated
63.3 cm
Aluminum, Epoxy, Polymer
57.9 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers