missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metro™ Starsys™ Preconfigured Mobile Workstation, LAR Imaging - Trauma Cart
Made from an easy-to-clean, advanced polymer material that will not dent, chip, rust, flake, or corrode like metal carts
Brand: Metro™ SXRSTRAMA
Additional Details : Weight : 92.89600kg
Includes: 45 in.H (113.6cm) Mobile shell with mechanical keylock and two keys, Three 3 in. (7.6cm) polymer drawers, Four 6 in. (15.2cm) polymer drawers, Two 3 in. drawer divider kits, Slate blue drawer pulls; Left swingout pod with top-locking tilt bin (keylock/two keys), Pod tray with dividers, Waste basket and holder; Right swingout pod with top-locking tilt bin (keylock/two keys), Middle nonlocking tilt bin, Sharps container bracket; Overbridge with shelf and rail, Utility/IV pole and clamp, Wire supply basket
Description
- Microban™ antimicrobial protection inhibits the growth of stain- and odor-causing bacteria
- Carts utilize 5 in. (127mm) polyurethane casters: two front casters with easily accessed toe brakes and two rear swivel casters
- Each drawer face has a 0.5 × 18.38 in. (1.3 × 46.7cm) polished area allowing for secure adhesion of DYMO™ and Zebra™ labels (not included)
Specifications
RTD Probe, 6 L x 0.18 in. dia., Stainless Steel, 4-wires Configuration, 100ohm(s) Precision, With 36 | |
44.7 in. | |
45 in. H Mobile shell with mechanical keylock and two keys, Three 3 in. Polymer drawers, four 6 in. Polymer drawers, Two 3 in. drawer divider kits, slate blue drawer pulls, left swingout pod with top-locking tilt bin (keylock/two keys), pod tray with dividers, waste basket and holder, Right swingout pod with top locking tilt bin (keylock/two keys), middle nonlocking tilt bin, sharps container bracket, overbridge with shelf and rail, utility/IV pole and clamp, wire supply basket | |
63.3 cm | |
34.7 in. |
24.9 x 34.7 x 44.7 in. (63.3 x 88.2 x 113.6 cm) | |
113.6 cm | |
24.9 in. | |
Polymer | |
88.2 cm |