missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metro™ Starsys™ Preconfigured Mobile Workstation, LAR Surgery Cart
Made from an easy-to-clean, advanced polymer material that will not dent, chip, rust, flake, or corrode like metal carts.
Brand: Metro™ SXRSANES
Includes: 45 in. (113.6 cm) mobile shell w/mechanical keylock and two keys, Four 6 in. (15.2 cm) polymer drawers, One 9 in. (22.9 cm) polymer drawer, Taupe drawer pulls, Left swing-out locking pod wi/three tilt bins (keylock w/two keys), Right swing-out locking pod w/three tilt bins (keylock w/two keys), Overbridge w/two rails; Universal clamp; Laryngoscope blade/specimen holder, Half-size metal utility bin, Full-size metal utility bin; Three label/tape dispensers, Six plastic utility bins w/clear covers
Description
- Advanced polymer based system with internal extruded aluminum structural support.
- Complete system of mobile and stationary units for storage, transportation and workstations that provide greater storage density in a smaller footprint.
- Single wide cart with left and right side swing-out accessory pods and overbridge with rails.
- Specifically designed to store and transport anesthesia supplies.
- Easily transport between stock room and procedure room.
- Swing out side pods provide additional work space.
- Each drawer face has a 0.5 x 18.38 in. (1.3 x 46.7 cm) polished area allowing for secure adhesion of DYMO™ and Zebra™ labels (not included.
Specifications
Anesthesia Cart | |
24.9 x 34.7 x 44.7 in. (63.3 x 88.2 x 113.6 cm) | |
44.7 in. | |
45 in. H mobile shell with mechanical keylock and two keys, four 6 in. polymer drawers, 9 in. polymer drawer, taupe drawer pulls, left swingout locking pod wi/three tilt bins (keylock with two keys), right swingout locking pod with three tilt bins (keylock with two keys), overbridge with two rails, universal clamp, laryngoscope blade/specimen holder, half size metal utility bin, full size metal utility bin, three label/tape dispensers, six plastic utility bins with clear covers | |
63.3 cm | |
None | |
88.2 cm |
Taupe | |
Storage and Transport | |
113.6 cm | |
24.9 in. | |
Polymer | |
34.7 in. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction