missing translation for 'onlineSavingsMsg'
Learn More

Metro™ Super Erecta™ Cart Handle

For customizing Super Erecta carts

Brand:  Metro™ H5S

Product Code. 15976931

  • £164.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • One piece
  • Stainless steel
  • 24 in. (61cm)
Specifications

Specifications

Cart Handle
Super Erecta Carts, Shelves and Casters
87.63 cm
24 in.
Damp environments like freezers, walk in coolers, laundry rooms and dishwashing stations
34.5 in.
Stainless Steel
60.96 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Metro™ Super Erecta™ Cart Handle > One-piece; Stainless-steel; 61cm (24 in.)

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.