missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metro™ Super Erecta™ SiteSelect™ Posts
For use with Metro Super Erecta, Super Adjustable Super Erecta and Super Erecta Pro wire shelving systems
Brand: Metro™ 86P
This item is not returnable.
View return policy
Description
Stationary posts are fitted with adjustable leveling bolts to compensate for uneven surfaces. Posts are available in Chrome, Type 304 Stainless Steel and Metroseal 3. Metroseal 3 includes Microban antimicrobial product protection and has a 12 year warranty against rust and corrosion. Mobile posts come without leveling bolt assembly for use with stem casters.
- Grooved at 1 in. (2.5cm) intervals
- Posts are double-grooved every 8 in. (20.3cm) for easy identification
Order Info
Height includes leveling bolt and cap. Shelves and posts sold separately.
Specifications
Metro™ Super Erecta™ wire shelving system | |
220 cm | |
Chrome |
86.63 in. | |
Stationary |