missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metro™ Super Erecta™ Wire Shelf, Chrome Finish
Wire shelves in chrome finish allow adjustable configurations when used with Metro Super Erecta shelving systems.
Brand: Metro™ 1824NC
This item is not returnable.
View return policy
Description
- Open-wire design minimizes dust accumulation, allows free circulation of air and greater visibility of stored items
- Real nickel-chrome finish for dry, low humidity environment
- Protective lacquer coated
Specifications
Wire Shelf | |
Metro™ Super Erecta™ shelving system | |
24 in. | |
800 lb. | |
18 in. | |
Chrome |
24 x 18 in. (61 x 45.7 cm) | |
Plastic sleeves and wedges | |
61 cm | |
363 kg | |
45.7 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction