missing translation for 'onlineSavingsMsg'
Learn More

Metro™ Super Erecta™ Wire Shelf, Stainless Steel Finish

Transform your laboratory or workplace storage as you see fit. Metro Super Erecta Wire Shelves in stainless steel finish allow adjustable configurations when used with Metro Super Erecta shelving systems

Brand:  Metro™ 1872NS

 View more versions of this product

Product Code. 15946611

  • £570.00 / Each
Estimated Shipment: 17-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Open-wire design minimizes dust accumulation, allows free circulatin of air and greater visibility of stored items
  • Type 304 polished stainless steel
  • Used for the most aggressive applications and environments
  • Stands up to high temperature automated cart wash and autoclave applications

Specifications

Specifications

18 in.
45.7 x 183 in. (18 x 72 cm)
Plastic sleeves and wedges
182.9 cm
45.7 cm
457 mm
Metro™ Super Erecta™ shelving system
72 in.
18 in.
Stainless Steel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers