missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metro™ Super Erecta™ Wire Shelf, Stainless Steel Finish
Transform your laboratory or workplace storage as you see fit. Metro Super Erecta Wire Shelves in stainless steel finish allow adjustable configurations when used with Metro Super Erecta shelving systems
Brand: Metro™ 1848NS
Additional Details : Weight : 4.44800kg
Description
- Open-wire design minimizes dust accumulation, allows free circulatin of air and greater visibility of stored items
- Type 304 polished stainless steel
- Used for the most aggressive applications and environments
- Stands up to high temperature automated cart wash and autoclave applications
Specifications
45.7 x 121.9 in. (18 x 48 cm) | |
18 in. | |
18 in. | |
45.7 cm | |
121.9 cm |
Plastic sleeves and wedges | |
48 in. | |
457 mm | |
Metro™ Super Erecta™ shelving system | |
Stainless Steel |