missing translation for 'onlineSavingsMsg'
Learn More

Metro™ MetroMax Shelf Posts

Compatible with MetroMax i™ and MetroMax Q™ shelves

Brand:  Metro™ MX33UP

 View more versions of this product

Product Code. 15917211

  • £53.45 / Each
Estimated Shipment: 27-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Grooves located every 1 in. for locking shelf wedges in place. Post height stamped to face of post for easy shelf positioning.

  • Corrosion-proof polymer posts with built-in Microban™ antimicrobial product protection
  • Stationary posts include an adjustable leveling foot to compensate for uneven floors
  • Leveling foot can be adjusted 1 in. (25mm)
  • Mobile posts accept optional Metro Stem Casters, available in a variety of materials to meet your specific needs

Specifications

Specifications

Mobile Post for Stem Caster
33 in.
Polymer
Metro™ MetroMax i™ and Q™ shelving
83.82 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers