missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metro™ MetroMax Shelf Posts
Compatible with MetroMax i™ and MetroMax Q™ shelves
Brand: Metro™ MX74P
Additional Details : Weight : 0.81600kg
Description
Grooves located every 1 in. for locking shelf wedges in place. Post height stamped to face of post for easy shelf positioning.
- Corrosion-proof polymer posts with built-in Microban™ antimicrobial product protection
- Stationary posts include an adjustable leveling foot to compensate for uneven floors
- Leveling foot can be adjusted 1 in. (25mm)
- Mobile posts accept optional Metro Stem Casters, available in a variety of materials to meet your specific needs
Specifications
Stationary Post with Leveling Foot | |
74 in. | |
Polymer |
188 cm | |
Metro™ MetroMax i™ and Q™ shelving |