Learn More
Mettler Toledo™ uPlace™ Electrode Holder for SevenCompact™ Meters
For use with SevenCompact S220 pH/Ion Meter and S230 Conductivity Meter.
Brand: Mettler Toledo™ 30019823
201.08 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : 03
Additional Details : Weight : 1.01700kg
Description
- uPlace electrode holder with the perfectly vertical uPlace movement helps you bring your electrode in the position where it works best for your sample
- This allows for faster measurements and reduces the risk of the sample vessel tipping over or damaging the sensor
Specifications
uPlace Electrode Holder | |
uPlace electrode holder with the perfectly vertical uPlace movement helps you bring your electrode in the position where it works best for your sample | |
SevenCompact S220 pH/Ion Meter and S230 Conductivity Meter |