Learn More
Mettler Toledo™ Adapters for pH Meters
Product Code. p-4677765 Shop All Mettler Toledo Products
Click to view available options
Type:
LTW-MiniDin Conductivity Adapter
Unit Size:
Each
Product Code. 11977731
Brand: Mettler Toledo™ 51302329
Only null left
Add to basket
Edge
Add to basket
Accessory adapters
LTW-MiniDin Conductivity Adapter
  • The appropriate cable to connect the conductivity sensor mini DIN to a portable meter.  Adapter is required for the following meters: FiveGo, Seven2Go, Seven2Go Pro, SevenGo Duo, SevenGo Duo Pro.
TRUSTED_SUSTAINABILITY
Specifications
Type LTW-MiniDin Conductivity Adapter
For Use With (Equipment) FiveGo, Seven2Go, Seven2Go Pro, SevenGo Duo, SevenGo Duo Pro