Learn More
Mettler Toledo™ Stoppers for Aquastar AQV33 Titrator
Replacement part for Aquastar AQV33 Titrator.
Brand: Mettler Toledo™ 00025883
Code : 04
Additional Details : Weight : 1.00000kg
Description
- For three-hole adapter
- 1 set (1 ST 10, 1 ST 7.5, 6 needle hole stoppers)
Specifications
Stopper |