Learn More
Mettler Toledo™ Balance Printer Accessories: Printer Paper
Printer paper for use with Mettler Toledo™ balances.
Brand: Mettler Toledo™ 72456
37.30 GBP valid until 2025-07-11
Use promo code "25721" to get your promotional price.
Description
- Color: White
- Five rolls per pack
- For use with Mettler Toledo balances: LC-P45, LC-P43, HA-P43, HR-73P

Specifications
LC-P45, LC-P43, HA-P43, HR-73P Printer | |
5 Rolls/Pk. | |
Printer Paper |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.