Learn More
METTLER TOLEDO™ InLab™ 413/IP67 pH Electrode
A durable, plastic body pH electrode ideal for a wide range of applications that is able to withstand wet and demanding environments (IP67 rated).
Brand: METTLER TOLEDO™ 52000379
349.99 GBP valid until 2025-07-11
Use promo code "25721" to get your promotional price.
Description
- For use with METTLER TOLEDO portable meters, SevenExcellence and SevenCompact as well as other brands. Connection cables may be required.
- Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
- Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.
Featured Applications
- Emulsions
- Suspensions
- Wastewater
- Effluent water
- Proteins & Sulfides
- General lab
Accessories & Care
- Open junction and Non-refillable - no additional electrolyte required
- Non-IP67 rated electrode available: InLab Expert Pro (51343101)
- Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
- Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)

Specifications
BNC/RCA IP67 | |
PEEK | |
0°C to 60°C | |
Polymer | |
Fixed Cable to sensor | |
InLab™ 413 | |
4.7 in. | |
120 mm | |
0.47 in. |
Open | |
0 to 14 | |
Combination Electrode; Integrated Temperature Probe | |
Ideal for emulsions, suspensions, effluent water and solutions containing Proteins and Sulphides | |
ARGENTHAL | |
0°F to 140°F | |
39 in. | |
1 m | |
12 mm |
Special Offers
- Multiple Manufacturers
-
Multiple Manufacturers
Dairy and Plant-Based Products Testing Promotion
Quality testing products for all your needs.Promo Code: 24134Expires: 2025-03-31
Days
0Hours
21Minutes
25Seconds
37 -
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Days
29Hours
2Minutes
34Seconds
22
Your input is important to us. Please complete this form to provide feedback related to the content on this product.