missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mettler Toledo™ Redox Buffer Solutions
Storage and refill solutions for redox electrodes.
Brand: Mettler Toledo™ 51350064
34.92 GBP valid until 2025-03-30
Use promo code "24134" to get your promotional price.
This item is not returnable.
View return policy
Description
Includes a quality inspection certificate which guarantees the stated values and traceability.Certifications
NIST traceable
Specifications
Yellow | |
Redox Buffer; UH/pH: 0.1 | |
468 mV | |
0.1 |
Buffer Solution | |
Quality inspection certificate | |
6 x 30 mL Bottle | |
Completely Miscible |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Redox buffer solution 468 mV pH 0.1
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Sulphuric acid
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Mettler Toledo™ Redox Buffer Solutions > UH/pH: 0.1; Packaging: 6 x 30mL bottle
Spot an opportunity for improvement?Share a Content Correction