missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mettler Toledo™ Redox Buffer Solutions
Shop All Mettler Toledo Products
Click to view available options
pH Range:
0.1
7
Filling Solution Type:
Redox Buffer; UH = 427mV
Redox Buffer; UH/pH: 0.1
Packaging:
250 mL Bottle
6 x 250 mL
6 x 30 mL Bottle
mV Range:
220 mV
468 mV
Unit Size:
1500mL
180mL
250mL
Description
Includes a quality inspection certificate which guarantees the stated values and traceability.
Certifications
NIST traceable
Specifications
Specifications
| Boiling Point | ∼100°C |
| Color | Clear, Yellow |
| Type | Buffer Solution |
| Filling Solution Type | Redox Buffer; UH = 427mV |
| Includes | Quality inspection certificate |
| mV Range | 220 mV |
| Packaging | 6 x 250 mL |
| pH Range | 7 |
| Water Soluble | Completely Miscible |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction