missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mettler Toledo™ Redox Buffer Solutions
Storage and refill solutions for redox electrodes.
£37.40 - £188.24
Specifications
Type | Buffer Solution |
---|---|
Includes | Quality inspection certificate |
Water Soluble | Completely Miscible |
Product Code | Brand | pH Range | Filling Solution Type | Packaging | mV Range | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | pH Range | Filling Solution Type | Packaging | mV Range | Price | Quantity & Availability | ||
10494071
|
Mettler Toledo™
51350060 |
7 | Redox Buffer; UH = 427mV | 250 mL Bottle | 220 mV |
£40.60
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12661306
|
Mettler Toledo™
51350062 |
7 | Redox Buffer; UH = 427mV | 6 x 250 mL | 220 mV |
£188.24
1500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10246082
|
Mettler Toledo™
51350064 |
0.1 | Redox Buffer; UH/pH: 0.1 | 6 x 30 mL Bottle | 468 mV |
£37.40
180mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
Includes a quality inspection certificate which guarantees the stated values and traceability.Certifications
NIST traceable
Specifications
Buffer Solution | |
Quality inspection certificate | |
Completely Miscible |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Mettler Toledo™ Redox Buffer Solutions