missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mettler Toledo™ Cap Kits and Solutions for Portable Dissolved Oxygen Meters
For use with Portable Dissolved Oxygen Meters.
£44.70
Specifications
For Use With (Equipment) | Portable Dissolved Oxygen Meter |
---|---|
Probe Type | DO |
Description
Kits and Solutions for use with Portable DO Meters.Specifications
Portable Dissolved Oxygen Meter | |
DO |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- zero oxygen tablets
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash hands thoroughly after handling.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : adipic acid
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title