Learn More
Mettler Toledo™ Connection Tube for Mettler Toledo Karl Fischer™ Titrators
Accessory for use with Mettler Toledo Karl Fischer Titrators.
Brand:  Mettler Toledo™ 00023957
Product Code. 12784802
  • £36.25 / Each
Estimated Shipment: 28-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Connects titration head to titration beaker drying tube
  • For model V20/30 Compact Karl Fischer titrator and DL31/38 Karl Fischer titrator
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Connection Tube