missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ Density Kit

For use with XS/XP balances. Mettler Toledo Density Kits helps to determine the density of solids and liquids with top-loading electronic balances.

Brand:  Mettler Toledo™ 11132680

Product Code. 10321560

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Solids Kit

  • 200mL beaker
  • Thermometer
  • Bridge
  • Sample Holder (2)
  • Bracket
  • Bottle of wetting agent, 25mL
Liquids Kit
  • Sinker mounted with pan and platinum wire

 

Note
Both kits are required to perform liquid density measurements.

Order Info

Made to order. Non-returnable.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

XS/XP Precision Balance
Solids kit includes 200mL beaker, thermometer, bri
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Mettler Toledo™ Density Kit > For XS/XP Precision balances (with 1mg and 0.1mg readability)

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.