missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ Electrode Fill Solutions

Product Code. p-3640208 Shop All Mettler Toledo Products
Click to view available options
Filling Solution Type:
1M KCl; 25 mL
FRISCOLYT-B
KCl saturated with AgCl; 25mL
For Use With (Equipment):
DX Series ISE Half Cell
Reference Electrode
Unit Size:
25mL
This item is not returnable. View return policy

Product Code. 11917781

Brand: Mettler Toledo™ 51343181

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Made from the highest quality reagents to provide optimum performance.

  • Aqueous solution containing inorganic and organic components
  • Supplied in 25mL (0.9 oz.) bottle

TRUSTED_SUSTAINABILITY

Specifications

Capacity (English) 0.9 oz.
Capacity (Metric) 25 mL
Concentration 1 mol/L
For Use With (Equipment) DX Series ISE Half Cell
Filling Solution Type 1M KCl; 25 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.