missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mettler Toledo™ Electrolytes for Reference Electrodes
Shop All Mettler Toledo Products
Click to view available options
Quantity:
250 mL
6 x 250 mL
Type:
FRISCOLYT-B
Unit Size:
1500mL
250mL
Description
- Electrolyte solution
- Contains ions, atoms or molecules that have lost or gained electrons, and is electrically conductive
Specifications
Specifications
| Description | Electrolyte FRISCOLYT-B, 6 x 250mL |
| Type | FRISCOLYT-B |
| Packaging | 6x 250 mL |
| Quantity | 6 x 250 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Mettler Toledo™ Electrolytes for Reference Electrodes >
Spot an opportunity for improvement?Share a Content Correction