missing translation for 'onlineSavingsMsg'
Learn More

METTLER TOLEDO™ InLab™ Viscous pH Electrodes

A glass pH electrode with an integrated temperature sensor featuring the SteadyForce™ reference system, which is pressurized to prevent the junction from clogging.

Brand:  METTLER TOLEDO™ 51343150

330.75 GBP valid until 2024-12-15
Use promo code "21658" to get your promotional price.


 View more versions of this product

Product Code. 12570137

  • £474.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
  • Intelligent Sensor Management (ISM) electrodes capture and save calibration data, which is then automatically recognized by the meter once connected.
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.

Featured Applications

  • Cosmetics
  • Gels, soaps, shampoos
  • Resins
  • Paints
  • Varnishes and glues
  • Colorants and dyes
  • Oily samples
  • TRIS compatible

Accessories & Care

  • Ceramic Junction and non-refillable - no additional electrolyte required
  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

pH Electrode InLab Viscous
Ceramic
0 to 14
Combination Electrode
Highly viscous, highly glutinous samples
SteadyForce
InLab™ Viscous
32°F to 266°F
1.57 in.
0.23 in.
S7
Glass
0°C to 130°C
FRISCOLYT-B
S7 to BNC (30281915)
HA Glass
3 mol/L KCl
0°C to 100°C
40 mm
6 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers