missing translation for 'onlineSavingsMsg'
Learn More

METTLER TOLEDO™ InLab™ Reference Electrodes

A reference electrode to be paired with half-cell measurement electrodes.

Brand:  METTLER TOLEDO™ 51343190

133.00 GBP valid until 2024-12-31
Use promo code "21658" to get your promotional price.


 View more versions of this product

Product Code. 10756513

  • £152.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.

Featured Applications

  • For use with half-cell measuring electrodes

Accessories & Care

  • S7 connector type: Cable S7 to 2 mm Banana (30281923) required
  • InLab Reference: ceramic junction
  • InLab Reference Plus: moveable PTFE sleeve junction
  • InLab Reference Flow: triple ceramic junction
  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

S7
Glass
Half-Cell Reference Electrode
For routine applications and measurements in Ion-selective half cells
InLab™ Reference
32°F to 212°F
4.72 in.
0.47 in.
Ceramic
0°C to 100°C
3mol/L KCl
ARGENTHAL
3mol/L KCl
0°C to 100°C
12 cm
1.2 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.