missing translation for 'onlineSavingsMsg'
Learn More

METTLER TOLEDO™ InLab™ Reference Electrodes

A reference electrode to be paired with half-cell measurement electrodes.

Brand:  METTLER TOLEDO™ 51343192

 View more versions of this product

Product Code. 10777304

  • £311.00 / Each
Estimated Shipment: 26-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.

Featured Applications

  • For use with half-cell measuring electrodes

Accessories & Care

  • S7 connector type: Cable S7 to 2 mm Banana (30281923) required
  • InLab Reference: ceramic junction
  • InLab Reference Plus: moveable PTFE sleeve junction
  • InLab Reference Flow: triple ceramic junction
  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

S7
Glass
Half-Cell Reference Electrode; Internal Gel Electrolyte
For routine applications and measurements in Ion-selective half cells
InLab™ Reference Flow
32°F to 266°F
4.72 in.
0.47 in.
Triple/Ceramic
0°C to 130°C
3mol/L KCl
ARGENTHAL
3mol/L KCl
0°C to 130°C
12 cm
1.2 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers