Learn More
Mettler Toledo™ Ion-Selective Combination Electrodes: Filling Solutions
For use with ISE combination electrodes.
Brand:  Mettler Toledo™ 51340030
32.70 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 12679185
  • £33.90 / 20mL
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
For use with ISE combination electrodes.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Electrolyte for Chloride ISE, 20mL
Ion-Selective Combination Electrodes Filling Solutions
Determine the Chloride concentration in samples
32°F to 176°F
20 mL
ISE Combination Electrode
1 mol/L KNO3
20 mL
0°C to 80°C
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Mettler Toledo™ Ion-Selective Combination Electrodes: Filling Solutions > Chloride Filling Solution