missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ Ion-Selective Combination Electrodes: Filling Solutions

Product Code. p-4883968 Shop All Mettler Toledo Products
Click to view available options
Filling Solution Type:
0.5 mol/L MgSO4
1 mol/L KNO3
3 mol/L KCl
For Use With (Equipment):
Ammonium ISE
Bromide ISE
Calcium ISE
Chloride ISE
Fluoride ISE
Nitrate ISE
Potassium ISE
Unit Size:
20mL
This item is not returnable. View return policy

Product Code. 12669185

Brand: Mettler Toledo™ 51340029

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For use with ISE combination electrodes.

For use with ISE combination electrodes.
TRUSTED_SUSTAINABILITY

Specifications

Description Electrolyte for Bromide ISE, 20mL
For Use With (Equipment) Bromide ISE
Type ISE Filling Solution
Filling Solution Type 1 mol/L KNO3
For Use With (Application) Determine the Bromide concentration in samples
Packaging Bottle
Quantity 20 mL
Temperature (English) Operating 32°F to 176°F
Temperature (Metric) Operating 0°C to 80°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.