missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ ISA Solutions

Solutions for perfectION™ combined ion selective electrodes

Brand:  Mettler Toledo™ 51344763

110.20 GBP valid until 2024-07-30
Use promo code "21657" to get your promotional price.



Code : 59

Additional Details : CAS Number : 7732-18-5 Weight : 0.58000kg

 View more versions of this product

Product Code. 11977511

  • £116.00 / 475mL
In Stock
to see stock.
Add to basket

Description

Description

Solutions (ionic-strength adjustment buffers)

  • Ion Electrolyte B (for chloride, copper, iodide, silver/sulfide ISE's), 475mL
  • ISA for calcium electrode, 475mL
  • ISA for potassium electrode, 475mL
  • ISA for nitrate electrode, 475mL
  • Nitrate ISS (for suppressing interference), 475mL
  • Fluoride TISAB II with CDTA, 3790mL
  • Fluoride TISAB III with CDTA (concentrate), 475mL

Specifications

Specifications

perfectION nitrate ISA
Nitrate ISA
475 mL
ISA Solution
475 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • perfectION Nitrate ISA

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3

Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH210-Safety data sheet available on request.

Precautionary Statement
  • P273-Avoid release to the environment.
  • P202-Do not handle until all safety precautions have been read and understood.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers