missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ ISA Solutions

Solutions for perfectION™ combined ion selective electrodes

Brand:  Mettler Toledo™ 51344764

Code : 59

Additional Details : CAS Number : 10043-35-3 Weight : 0.53000kg


 View more versions of this product

Product Code. 11987511

  • £112.00 / 475mL
In Stock
to see stock.
Add to basket

Description

Description

Solutions (ionic-strength adjustment buffers)

  • Ion Electrolyte B (for chloride, copper, iodide, silver/sulfide ISE's), 475mL
  • ISA for calcium electrode, 475mL
  • ISA for potassium electrode, 475mL
  • ISA for nitrate electrode, 475mL
  • Nitrate ISS (for suppressing interference), 475mL
  • Fluoride TISAB II with CDTA, 3790mL
  • Fluoride TISAB III with CDTA (concentrate), 475mL

Specifications

Specifications

perfectION nitrate ISS
Nitrate ISS
475 mL
ISA Solution
475 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P202-Do not handle until all safety precautions have been read and understood.
  • P273-Avoid release to the environment.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers