Learn More
Mettler Toledo™ Mettler Toledo™ Micro pH Electrode: LE422
Designed for measuring in small or narrow neck vessels or test tubes
Brand: Mettler Toledo™ 30089747
196.08 GBP valid until 2024-07-30
Use promo code "21657" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.11700kg
Description
- Ag/AgCl reference system
- Gel electrolyte filling
- Low maintenance
- U-membrane glass
- Glass shaft
- Ceramic frit
- LE cap at electrode head
- pH Range: 0 to 14
- Zero Point (pH): 7.00±0.25
- Temperature Range: 0° to 80°C
- Membrane Resistance: <250MOhm
- Lower Shaft Length: 90mm
- Lower Shaft Dia.: 4.3mm
- Cable Length: 1m
- Connector: BNC
Recommended Storage
Store for short term in 3mol/L KCl in watering cap and for long-term storage, store in 3mol/L KCl, buffer pH 4 or 7.
Specifications
pH electrode LE422 | |
Ag/AgCl | |
0 to 14 | |
Measuring in small or narrow neck vessels or test tubes. | |
Store for short term in 3mol/L KCl in watering cap and for long-term storage store in 3mol/L KCl, buffer pH 4 or 7. | |
1 m |
BNC | |
Glass | |
Gel | |
pH electrode LE422 | |
0°C to 80°C | |
90 mm |