missing translation for 'onlineSavingsMsg'
Learn More
Learn More
METTLER TOLEDO™ InLab™ Redox ORP Electrodes
An ORP/Redox electrode featuring a metal ring indicator designed for measuring oxidation/reduction potential; ideal for water and other general applications.
£252.00 - £433.00
Specifications
Connector Types | S7 |
---|---|
Material | Glass |
For Use With (Application) | For measuring the oxidation/reduction power of solution |
For Use With (Equipment) | S7 to BNC (30281915) |
Product Code | Brand | Model | Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Type | Price | Quantity & Availability | ||||
10786733
|
METTLER TOLEDO™
51343200 |
InLab™ Redox | Combination Electrode; Platinum ring indicator |
£316.00
Each |
In Stock
Log in to see stock. |
||||
10683004
|
METTLER TOLEDO™
51343205 |
InLab™ Redox Ag | Combination Electrode; Silver ring indicator |
£329.00
Each |
In Stock
Log in to see stock. |
||||
10735583
|
METTLER TOLEDO™
51343204 |
InLab™ Redox Au | Combination Electrode; Gold ring indicator |
£419.00
Each |
In Stock
Log in to see stock. |
||||
10028142
|
METTLER TOLEDO™
51343201 |
InLab™ Redox Flow | Combination Electrode; Platinum ring indicator |
£433.00
Each |
In Stock
Log in to see stock. |
||||
10531645
|
METTLER TOLEDO™
51343203 |
InLab™ Redox Micro | Combination Electrode; Platinum ring indicator |
£330.00
Each |
Estimated Shipment: 04-02-2025
Log in to see stock. |
||||
10353713
|
METTLER TOLEDO™
51343202 |
InLab™ Redox-L | Combination Electrode; Platinum ring indicator |
£322.00
Each |
In Stock
Log in to see stock. |
||||
10732722
|
METTLER TOLEDO™
59904377 |
InLab™ Redox Pt805 | Half-Cell Electrode; Platinum ring indicator |
£252.00
Each |
Estimated Shipment: 04-02-2025
Log in to see stock. |
||||
10748972
|
METTLER TOLEDO™
59904391 |
InLab™ Redox Ag805 | Half-Cell Electrode; Silver ring indicator |
£289.00
Each |
Estimated Shipment: 04-02-2025
Log in to see stock. |
||||
Description
- For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
- Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
- Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.
Featured Applications
- Measuring oxidation/reduction potential (ORP)
- Drinking water
- Cooling towers
- Swimming pools
- Potable water supplies
- Waste water
- General lab
Accessories & Care
- Ceramic junction (excluding the InLab Redox Flow with movable glass sleeve junction)
- Refillable 3M KCl (51350072): Keep probe filled with electrolyte for accurate results
- Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
- InLab Redox Pt805 and InLab Redox Ag805: no electrolyte required and must be paired with InLab Reference (51343190)
- Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
Specifications
S7 | |
Glass | |
For measuring the oxidation/reduction power of solution | |
S7 to BNC (30281915) |