Learn More
Mettler Toledo™ Replacement Parts for V20 Karl Fischer™ Autotitrator
For use with V20 Karl Fischer Autotitrators.
Brand:  Mettler Toledo™ 00023961
 View more versions of this product
Product Code. 11351590
  • £140.00 / Each
Estimated Shipment: 09-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Accessory for DV1000 titration burettes
  • For titrant protection from moisture or CO2
  • With cover
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Drying Cup
Water content determination
453.5 g