missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
METTLER TOLEDO™ SevenExcellence™ S400 Benchtop pH Meter
Handla allt Mettler Toledo Produkter
Klicka för att se tillgängliga alternativ
Model:
Meter, Bio/Life Sciences Kit
Meter, Standard Kit
S400 pH Meter
Förpackningsstorlek:
Each
Beskrivning
- pH, Redox/ORP, and ISFET meter than can be expanded with additional measurement modules to measure conductivity, resistivity, dissolved oxygen, ISE, salinity, TDS, and BOD
- Easily create measurement and calibration shortcuts
- 7 inch color touch screen with intuitive menu navigation
- Compatible with a wide range of electrodes for any application
- *Compatible with our ISM electrodes that capture and save calibration data which is then automatically recognized by the meter once connected
- Meter connects to electrodes via BNC; S7 to BNC cables available
- Export data to a printer, Excel file, or LIMS system
- Automation available for measurements and cleaning in high-throughput applications
- Manage user permissions and access to ensure data integrity
- Compatible with LabX software for advanced data management and 21CFR part 11 regulatory compliance
- Optional comprehensive qualification package available including IQ/OQ/PQ
- Preventative maintenance and service available through METTLER TOLEDO Service
Specifikationer
Specifikationer
| Includes | S400 pH meter; electrode holder; 2 buffer sachets for pH 4.01, 7, 9.21 and 10.01; EasyDirect pH activation key (pH data management software); test certificate |
| Accuracy (Temperature) | ±0.1°C |
| Accuracy (mV) | ±0.1 |
| Accuracy (pH) | ±0.002 |
| Outputs | USB, RS-232, Ethernet |
| Range (pH) | -2 to 20 |
| Resolution (pH) | 0.001, 0.01, 0.1 |
| Calibration Points (pH) | Max. 5 points, 8 pre-defined and 20 user-defined buffer group |
| Range (mV) | -2000 to 2000 |
| Resolution (mV) | 0.1/1mV |
| Visa mer |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering