Learn More
Mettler Toledo™ SevenGo™ Dissolved Oxygen Replacement Electrode InLab™ 605
Accessory for use with InLab 605 Dissolved Oxygen Electrode.
Brand: Mettler Toledo™ 51340293
233.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Includes: Three membrane bodies and 25mL of electrolyte
Description
For use with InLab™ 605 Dissolved Oxygen Electrode.
Specifications
Membrane kit InLab 605 | |
Three membrane bodies and 25mL of electrolyte | |
InLab 605 Dissolved Oxygen Electrode | |
Routine pH measurement |
Safety and Handling
- MEMBRANE KIT INLAB 605
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H319-Causes serious eye irritation.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Potassium hydroxide;caustic potash
Special Offers
- Multiple Manufacturers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Your input is important to us. Please complete this form to provide feedback related to the content on this product.