Learn More
Mettler Toledo™ V20 Three-Hole Adapter for Titration Stand
Three-hole adapter for titration stand.
Brand:  Mettler Toledo™ 00023982
Product Code. 10386408
  • £69.21 / Each
Estimated Shipment: 10-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For use with V20 and V30 Karl Fischer titrators
  • Includes ST10, ST7.5 and needle hole and pin hole stoppers
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Adapter
Standard 10 and 7.5 needle hole and appropriate stopper
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Mettler Toledo™ V20 Three-Hole Adapter for Titration Stand > Three-hole adapter